Anti-GALR2 Rabbit polyclonal antibody
ORIGTA308347
: OriGene
- Antibody type:Primary
- Antigen name:Galanin Receptor 2
- Antigen symbol:GALR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Form:Liquid
- Amino acid number:212 to 307
- Storage buffer:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7). 0,01% Thimerosal was added as a preservative.
- Molecular weight:42 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by antigen affinity chromatography
- Pk:100 µl
Recommended Dilutions: Immunochemistry1:100-1:1000; Western Blot:1:500-1:3000
Type: Primary
Antigen: GALR2 (galanin receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human