Order Entry
Denmark
ContactUsLinkComponent
Anti-E2-Crimson Mouse Monoclonal Antibody
Anti-E2-Crimson Mouse Monoclonal Antibody
Catalog # ORIGTA180085
Supplier:  OriGene
Anti-E2-Crimson Mouse Monoclonal Antibody
Catalog # ORIGTA180085
Supplier:  OriGene

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    E2-Crimson
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µl

Specifications

About this item

Type: Tag
Antigen: E2-Crimson
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: