Order Entry
Denmark
ContactUsLinkComponent
 
Anti-PLVAP Rabbit Polyclonal Antibody
  ABGEAP9378C
 :  Abgent
undefined
Anti-PLVAP Rabbit Polyclonal Antibody
  ABGEAP9378C
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Plasmalemma Vesicle Associated Protein
  • Antigen symbol:
    PLVAP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-Terminal
  • Gene ID:
    226778
  • Antigen synonyms:
    Serine/threonine-protein kinase p78|ELKL motif kinase 1|Par1c|EMK-2|CTAK1|EMK-1|PAR1 homolog c|cTAK1|MARK3|MARK|Serine/threonine-protein kinase MARK1|MARK4|PAR1 homolog|Par1b|Protein kinase STK10|MAP/microtubule affinity-regulating kinase 4|PAR1 homolog b|MAP/microtubule affinity-regulating kinase 3|Ser/Thr protein kinase PAR-1|MARKL1|KIAA1860|C-TAK1|EMK1|Par-1a|MARK1|EMK2|MAP/microtubule affinity-regulating kinase 1|Serine/threonine-protein kinase MARK2|MAP/microtubule affinity-regulating kinase 2|Cdc25C-associated protein kinase 1|KIAA1477|Par-1c|MARK2|MAP/microtubule affinity-regulating kinase-like 1|ELKL motif kinase 2|Par-1b
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Sequence:
    HGEQLRERRSAAYC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This Mouse Mark1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 600-627 amino acids from the C-terminal region of mouse Mark1.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: PLVAP (plasmalemma vesicle associated protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human