Anti-GABRD Rabbit Polyclonal Antibody
ABGEAP9299C
: Abgent
- Antibody type:Primary
- Antigen name:Gamma-aminobutyric acid (GABA) A Receptor delta
- Antigen symbol:GABRD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Gene ID:54623
- Antigen synonyms:Gamma aminobutyric acid receptor delta subunit precursor GABA A receptor|Gamma aminobutyric acid GABA A receptor delta|MGC45284.|Gamma-aminobutyric acid receptor subunit delta|GBRD_HUMAN|GABRD|GABAA receptor subunit delta
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:FGSDADSEDDADSDC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,4 mg/ml
- Shipping temperature:4 °C
- Immunogen:This PAF1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 439-467 amino acids from the C-terminal region of human PAF1.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: GABRD (gamma-aminobutyric acid (GABA) A receptor, delta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human