Anti-CREM Rabbit Polyclonal Antibody
ABGEAP9122B
: Abgent
- Antibody type:Primary
- Antigen name:Discoidin domain receptor family member 2
- Antigen symbol:DDR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Gene ID:18214
- Antigen synonyms:Receptor protein-tyrosine kinase TKT|receptor-related 3|Neurotrophic tyrosine kinase|NTRKR3|DDR2|Discoidin domain receptor 2|TKT|Tyrosine-protein kinase TYRO 10
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:WRRETKHRPSFQEC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This Mouse Ddr2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 816-843 amino acids from the C-terminal region of mouse Ddr2.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: CREM (cAMP responsive element modulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human