Order Entry
Denmark
ContactUsLinkComponent
 
Anti-LRPAP1 Rabbit Polyclonal Antibody
  ABGEAP8529B
 :  Abgent
undefined
Anti-LRPAP1 Rabbit Polyclonal Antibody
  ABGEAP8529B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Low density lipoprotein receptor-related protein associated protein 1
  • Antigen symbol:
    LRPAP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-Terminal
  • Gene ID:
    11338
  • Antigen synonyms:
    MRAP|HBP44|A2MRAP|A2RAP|RAP|MYP23|alpha-2-MRAP
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Sequence:
    FERQLNENKQERDKC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This U2AF2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human U2AF2.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: LRPAP1 (low density lipoprotein receptor-related protein associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human