Anti-TK2 Rabbit Polyclonal Antibody
ABGEAP7062A
: Abgent
- Antibody type:Primary
- Antigen name:Thymidine kinase 2
- Antigen symbol:TK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-Terminal (1–100 aa)
- Cross adsorption:No
- Antigen synonyms:mitochondrial|Thymidine kinase 2|Mt-TK
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: TK2 (thymidine kinase 2, mitochondrial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse