Anti-TLE4 Rabbit Polyclonal Antibody
Catalog # ABGEAP12953A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Transducin-like enhancer of split 4
- Antigen symbol:TLE4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Antigen synonyms:ESG4|Grg-4|ESG|BCE1|GRG4|BCE-1|E(spI)
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: TLE4 (transducin-like enhancer of split 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human