Anti-HIST1H2AE Rabbit Polyclonal Antibody
AVIVARP70539_P050
- Antibody type:Primary
- Antigen name:histone cluster 1, H2ae
- Antigen symbol:HIST1H2AE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:3012
- Antigen synonyms:H2A.1|H2A/a|H2AFA|H2A.2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: HIST1H2AE (histone cluster 1, H2ae)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human