Anti-EFTUD2 Rabbit Polyclonal Antibody
Catalog # AVIVARP70182_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:elongation factor Tu GTP binding domain containing 2
- Antigen symbol:EFTUD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:9343
- Antigen synonyms:SNRNP116|Snrp116|MFDGA|MFDM|Snu114|U5-116KD
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: EFTUD2 (elongation factor Tu GTP binding domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human