Anti-PDZD4 Rabbit Polyclonal Antibody
AVIVARP68784_P050
- Antibody type:Primary
- Antigen name:PDZ Domain Containing 4
- Antigen symbol:PDZD4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:57595
- Antigen synonyms:LU1|PDZRN4L|PDZK4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: PDZD4 (PDZ domain containing 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human