Anti-MARC2 Rabbit Polyclonal Antibody
Catalog # AVIVARP68677_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Membrane associated ring-CH-type finger 2
- Antigen symbol:MARCH2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:54996
- Antigen synonyms:RNF172|MARCH-II|HSPC240
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: MARC2 (mitochondrial amidoxime reducing component 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human