Anti-KRT31 Rabbit Polyclonal Antibody
AVIVARP67626_P050
- Antibody type:Primary
- Antigen name:keratin 31, type I
- Antigen symbol:KRT31
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:3881
- Antigen synonyms:HA1|KRTHA1|Ha-1|hHa1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: KRT31 (keratin 31)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human