Anti-RAB6C Rabbit Polyclonal Antibody
Catalog # AVIVARP60501_P050
Supplier: Aviva Systems Biology
Jump to:
Product Details & Documents
Type: Primary
Antigen: RAB6C (RAB6C, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Documents
ARP60501_P050.pdf
Datasheet
Specifications
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- Epitope:middle
- Storage temperature:Cool
- Host:Rabbit
- Cross adsorption:none
- Shipping temperature:Ice
- Antigen name:RAB6C, member RAS oncogene family
- Cat. No.:AVIVARP60501_P050
- Reactivity:Human
- Gene ID:84084
- Antigen synonyms:WTH3
- Conjugation:Unconjugated
- Antigen symbol:RAB6C
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



