Anti-PRRX2 Rabbit Polyclonal Antibody
AVIVARP57884_P050
- Antibody type:Primary
- Antigen name:Paired related homeobox 2
- Antigen symbol:PRRX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Pig,Human,Bovine,Dog,Rat
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:51450
- Antigen synonyms:PMX2|PRX2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: PRRX2 (paired related homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Pig, Rat