Anti-PLEKHB2 Rabbit Polyclonal Antibody
Catalog # AVIVARP56309_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Pleckstrin homology domain containing B1
- Antigen symbol:PLEKHB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:55041
- Antigen synonyms:EVT2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: PLEKHB2 (pleckstrin homology domain containing, family B (evectins) member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Rat