Anti-PIGV Rabbit Polyclonal Antibody
AVIVARP47358_T100
- Antistof type:Primary
- Antigen name:phosphatidylinositol glycan anchor biosynthesis, class V
- Antigen symbol:PIGV
- Clonality:Polyclonal
- Konjugation:Unconjugated
- Vært:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitop:N-terminal
- Krydsadsorption:none
- Gen ID:55650
- Antigen synonymer:PIG-V|GPI-MT-II|HPMRS1
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Pakn:100 µG
Type: Primary
Antigen: PIGV (phosphatidylinositol glycan anchor biosynthesis, class V)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: