Anti-NOX1 Rabbit Polyclonal Antibody
Catalog # AVIVARP46818_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:NADPH oxidase 1
- Antigen symbol:NOX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:27035
- Antigen synonyms:NOH-1|MOX1|NOH1|GP91-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: NOX1 (NADPH oxidase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: