Anti-B3GALNT1 Rabbit Polyclonal Antibody
Catalog # AVIVARP44858_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:beta-1,3-N-acetylgalactosaminyltransferase 1 (globoside blood group)
- Antigen symbol:B3GALNT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:8706
- Antigen synonyms:GLOB|Gb4Cer|P|P1|galT3|B3GALT3|GLCT3|beta3Gal-T3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: B3GALNT1 (beta-1,3-N-acetylgalactosaminyltransferase 1 (globoside blood group))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: