Anti-PON1 Rabbit Polyclonal Antibody
AVIVARP41400_P050
- Antibody type:Primary
- Antigen name:paraoxonase 1
- Antigen symbol:PON1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:5444
- Antigen synonyms:PON|ESA|MVCD5
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: PON1 (paraoxonase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: