Anti-RPL6 Rabbit Polyclonal Antibody
AVIVARP40213_T100
- Antibody type:Primary
- Antigen name:Ribosomal Protein L6
- Antigen symbol:RPL6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:6128
- Antigen synonyms:SHUJUN-2|TAXREB107|TXREB1|L6
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: RPL6 (ribosomal protein L6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Rat