Anti-AHRR Rabbit Polyclonal Antibody
Catalog # AVIVARP39438_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:aryl-hydrocarbon receptor repressor
- Antigen symbol:AHRR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:57491
- Antigen synonyms:AHHR|AHH|bHLHe77
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: AHRR (aryl-hydrocarbon receptor repressor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: