Order Entry
Denmark
ContactUsLinkComponent
 
Anti-MED21 Rabbit Polyclonal Antibody
  AVIVARP35728_P050
Anti-MED21 Rabbit Polyclonal Antibody
  AVIVARP35728_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    Mediator Complex Subunit 21
  • Antigen symbol:
    MED21
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    108098
  • Antigen synonyms:
    D19234|D6Ertd782e|0610007L03Rik|Surb7|AI449604|Srb7
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: MED21 (mediator complex subunit 21)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: