Anti-PITX2 Rabbit Polyclonal Antibody
Catalog # AVIVARP32431_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Paired-like homeodomain 2
- Antigen symbol:PITX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:5308
- Antigen synonyms:ARP1, Brx1, IDG2, IGDS, IGDS2, IHG2, IRID2, Otlx2, PTX2, RGS, RIEG, RIEG1, RS
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: PITX2 (paired-like homeodomain 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: