Order Entry
Denmark
ContactUsLinkComponent
Anti-RBM10 Rabbit Polyclonal Antibody
Anti-RBM10 Rabbit Polyclonal Antibody
Catalog # AVIVARP30104_T100
Anti-RBM10 Rabbit Polyclonal Antibody
Catalog # AVIVARP30104_T100

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    RNA binding motif protein 10
  • Antigen symbol:
    RBM10
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    8241
  • Antigen synonyms:
    S1-1|TARPS|ZRANB5|GPATCH9|DXS8237E|GPATC9
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: RBM10 (RNA binding motif protein 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: