Anti-TSPYL2 Rabbit Polyclonal Antibody
BOSSBS-7051R
: Bioss
- Antibody type:Primary
- Antigen name:TSPY-like 2
- Antigen symbol:TSPYL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Antigen synonyms:Testis-specific Y-encoded-like protein 2|Cutaneous T-cell lymphoma-associated antigen se20-4|DENTT|TSPX|Cell division autoantigen 1|Testis-specific Y-enc|CDA1|TSPYL2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Type: Primary
Antigen: TSPYL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat