Anti-mu Opioid receptor Rabbit Polyclonal Antibody
BOSSBS-3623R
: Bioss
- Antibody type:Primary
- Antigen name:Mu Opioid Receptor
- Antigen symbol:OPRM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Antigen synonyms:OPRM1|MOR-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Type: Primary
Antigen: mu Opioid receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat