Anti-TRAF2 Rabbit Polyclonal Antibody
BOSSBS-1213R
: Bioss
- Antibody type:Primary
- Antigen name:TNF receptor-associated factor 2
- Antigen symbol:TRAF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Antigen synonyms:TRAP|TRAP3|TNF receptor-associated factor 2|TRAF2|MGC,45012|E3 ubiquitin-protein ligase TRAF2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Type: Primary
Antigen: TRAF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat