HPLC columns, SUPELCOSIL™, LC-18, Supelco®
Supplier: Merck
Certificates
About this item
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Low TOC cleaning validation swabs, Alpha®
Supplier: Texwipe
VWR®, Nitrile Gloves
Supplier: VWR Collection
HPLC columns, LiChrosorb®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Single use syringes, 2-piece, Injekt® Solo
Supplier: B. Braun
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
HPLC columns, LiChrospher®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Screw caps for digestion tubes, DigiTUBE®
Supplier: PerkinElmer