Order Entry
Denmark
ContactUsLinkComponent
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
Catalog # 786-514
Supplier:  G-Biosciences
CAS Number:  
undefined
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
Catalog # 786-514
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol
 :
UN:3082
CAS Number:9002-93-1
 :9036-19-5
MDL Number:MFCD00132505
Melting Pt:6
Flash Pt:251
ADR:9 III
Merck Index:13,06793