Medical tubing, Tygon® ND 100-65
Supplier: Saint Gobain Life Sciences
Certificates
About this item
Tygon®, crystal clear.
- Crystal clear for easy visualisation of fluid flow
- Ideal for contact with bodily fluids
- Non-wetting surface permits complete drainage
- Meets USP Class VI criteria
- REACH compliant
- Withstands EtO, gas and gamma sterilisation
Developed specifically for use in medical applications.
Temperature resistance: –42 to +74 °C
Hardness (Shore durometer A): 65
Density: 1,19
Frequently Bought Together