PCR plates, 384-well
Catalog # 391-5711
Supplier: Brand
Specifications
- No. of wells:384
- Profile:Low
- Skirt:Fully skirted
- Well colour:Clear
- Description:PCR plate (cut corners A12, H12)
- Environmentally Preferable:
- Pk:50
- Well volume:0,04 ml
Specifications
About this item
PP. The 40 μl wells can hold sample volumes of 2 to 30 μl. This helps to reduce reagent costs and shortens cycle times. The plates can be filled using multichannel pipettes or robotic systems.
- Ultra-thin walled design to facilitate constant, rapid and precise heat transfer
- Smooth vessel interior minimises the binding of enzymes and nucleic acid to the walls
- Embossed alphanumeric coding
- Raised well rims protect against cross-contamination and provide a tight seal in combination with matching sealing mat
- Free from DNase, RNase and DNA
- Autoclavable at 121 °C (2 bar) acc. DIN EN 285