10-Bromodecanoic acid ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:50530-12-6
:
Formula:C₁₀H₁₉BrO₂
MDL Number:MFCD00014388
Molecular Weight:251.16 g/mol
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light yellow to Green powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:39.0 to 45.0 deg-C
Specification Test Results
Frequently Bought Together