trans-Bis(tributylstannyl)ethylene 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: trans-1,2-Bis(tri-n-butylstannyl)ethylene
CAS Number:14275-61-7
:
Formula:C₂₆H₅₆Sn₂
MDL Number:MFCD01631299
Molecular Weight:606.15 g/mol
Storage Temperature:Refrigerator
UN:2788
ADR:6.1 III
:GHS06
:GHS08
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5000 to 1.5030 (20°C, 589 nm)
- NMR:≥94 %
Specification Test Results
Frequently Bought Together