Order Entry
Denmark
ContactUsLinkComponent
Diode laser, red
  DIDAPOD013135
 :  DIDALAB
undefined
Diode laser, red
  DIDAPOD013135
 :  DIDALAB
  • Description:
    Diode laser, red
  • Environmentally Preferable:
  • Pk:
    1
  • Wavelength:
    650 nm
The diameter of the beam is adjustable from 1 to 12 mm.
 : Supplied with Ø 10-mm rod and mains adapter.