Order Entry
Denmark
ContactUsLinkComponent
 
Sodium thiosulphate 0.039 mol/l (0.039 N) in aqueous solution 0.039 mol/l (0.039 N) in aqueous solution
Catalog # 88946.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium thiosulphate 0.039 mol/l (0.039 N) in aqueous solution 0.039 mol/l (0.039 N) in aqueous solution
Catalog # 88946.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
 :
 :158.11 g/mol
Boiling Pt:100
Melting Pt:48
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499
Merck Index:12,08844