Order Entry
Denmark
ContactUsLinkComponent
 
1-(2-Aminoethyl)piperidine 98%
 
undefined
1-(2-Aminoethyl)piperidine 98%

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4730 to 1.4750 (20°C, 589 nm)
  • Titration with HClO4:
    ≥97.5 %

 

 

Frequently Bought Together