Heat and cold resistant gloves with chemical protection, Tricotril® Winter
These gloves are resistant to a variety of chemicals and provide short-term protection against cold down to −20 °C and against heat up to 250 °C. The heavy seamless wool liners are fully nitrile coated.
- Broad chemical resistance, high mechanical resistance
- Textured profile for good grip when handling greasy and oily parts
- Free from natural latex
- Good antistatic and electrostatic properties
- Long cuff also protects forearm
Ideal applications include working in cold areas and environments with steam hazards, handling of contaminated heavy parts, cleaning and maintenance in chemical plants, galvanising works and chemical industry applications.
Thickness: 2,0 ± 0,2 mm
Colour: Green
EN 388 (3121), EN 374 (AJL), EN 407 (X1XXXX), Cat. III Food contact approved
: EN 388:2016 (3121X), EN ISO 374-1 Type A (AJKLOPT), EN 407 (X1XXXX), Cat. III
Food contact approved
Food contact approved
- Catalog No:
- KCHL739.9
- KCHL739.8
- KCHL739.11
- KCHL739.10
- Model:
- 737
- 737
- 737
- 737
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Wool
- Wool
- Wool
- Wool
- Finish:
- Textured palm
- Textured palm
- Textured palm
- Textured palm
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- EN norm:
- EN 388:2016 (3121X), EN ISO 374-1 Type A (AJKLOPT), EN 407 (X1XXXX)
- EN 388:2016 (3121X), EN ISO 374-1 Type A (AJKLOPT), EN 407 (X1XXXX)
- EN 388:2016 (3121X), EN ISO 374-1 Type A (AJKLOPT), EN 407 (X1XXXX)
- EN 388:2016 (3121X), EN ISO 374-1 Type A (AJKLOPT), EN 407 (X1XXXX)
- Anti-static:
- +
- +
- +
- +
- Food contact approved:
- +
- +
- +
- +
Frequently Bought Together