Bulkhead unions, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Certificates
About this item
Designed for plumbing tubing through equipment housing. Each union has unique ³/₈-24 external threads and comes complete with a stainless steel nut and lock washer to hold it in place.
- Requires a ³/₈" hole to mount
- Recommended torque limit 15 in.– lbs (1,7 N-m)
Frequently Bought Together