Order Entry
Denmark
ContactUsLinkComponent
≥98.0% (by titrimetric analysis) good's buffer component for biological research
≥98.0% (by titrimetric analysis) good's buffer component for biological research
Catalog # TCIAH0576-25G
Supplier:  TCI
CAS Number:  
undefined
≥98.0% (by titrimetric analysis) good's buffer component for biological research
Catalog # TCIAH0576-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    min. 98.0 %
  • Absorbance:
    max. 0.15 (0.1mol/L、H2O、260nm)
  • Solubility in Water:
    almost transparency
  • Heavy metals (as Pb):
    max. 5 ppm

Specification Test Results

About this item

Synonyms: 4-(2-Hydroxyethyl)-1-piperazinepropanesulphonic acid, N-(2-Hydroxyethyl)piperazine-N′-(3-propanesulphonic acid), HEPPS, 3-[4-(2-Hydroxyethyl)-1-piperazinyl]propanesulphonic acid, 3-[4-(2-Hydroxyethyl)-1-piperazinyl]-propanesulphonic acid, 4-(2-Hydroxyethyl)piperazine-1-propanesulphonic acid
 :
 :252.33 g/mol
EINECS:240-198-8
 :GHS07
Storage Temperature:Ambient
CAS Number:16052-06-5
Formula:C₉H₂₀N₂O₄S
MDL Number:MFCD00006160