(R)-4-Benzyloxy-1,2-butanediol ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:86990-91-2
:
Formula:C₁₁H₁₆O₃
MDL Number:MFCD10000949
Boiling Pt:168°C
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
- Specific rotation [a]20/D:+21.0 to +25.0 deg (C=5, EtOH)
Specification Test Results
Frequently Bought Together