Capillary columns, TraceGOLD™ TG-WaxMS
TraceGOLD™ TG-WaxMS columns are manufactured for better column-to-column reproducibility.
- Polar phase, polyethylene glycol
- Polar-deactivated surface tightly binds polymer for excellent thermal stability
- Resists oxidative damage, damage from strongly acidic or basic volatiles better than silicone solid phases
- Equivalent to USP G14, G15, G16, G20 and G39 phases
Frequently Bought Together