Order Entry
Denmark
ContactUsLinkComponent
 
Protein A
Catalog # 17-0872-50
Supplier:  Cytiva
CAS Number:  
undefined
Protein A
Catalog # 17-0872-50
Supplier:  Cytiva
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Source:
  • Species:
  • Environmentally Preferable:
  • Form:
    Freeze-dried
  • Protein/peptide name:
    Protein A

Specifications

About this item

Staphylococcal protein A is the classical immunoglobulin-binding reagent for a wide range of antibodies, used extensively in affinity chromatography and antibody detection applications. The protein is enterotoxin-free and gives reliable results. Protein A binds the Fc regions of IgG antibodies from a variety of species, for convenient antibody purification.