Order Entry
Denmark
ContactUsLinkComponent
Cation exchange chromatography columns, HiTrap™ Capto™ S
Cation exchange chromatography columns, HiTrap™ Capto™ S
Supplier:  Cytiva
Certificates
undefined
Cation exchange chromatography columns, HiTrap™ Capto™ S
Supplier:  Cytiva
Specifications
  • Catalog No:
  • 29-4004-58
  • 17-5441-22
  • 17-5441-23
  • Max. pressure:
  • 5 bar
  • 5 bar
  • 5 bar
  • pH range:
  • 4 - 12
  • 4 - 12
  • 4 - 12