2-Fluoro-4-nitroaniline ≥98.0% (by GC)
Leverandør: TCI
Certifikater
Om denne vare
CAS nummer:369-35-7
:
Formel:C₆H₅FN₂O₂
MDL nummer:MFCD00034560
Molecular Weight:156.12 g/mol
EINECS:206-719-8
:GHS07
Opbevaringstemperatur:Ambient
Specifikationer
- Environmentally Preferable:
Specifikationer
Specifikationstestresultater
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:133.0 to 137.0 deg-C
Specifikationstestresultater
Frequently Bought Together