1,4-Dibromo-2,5-bis(bromomethyl)benzene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:35335-16-1
:
Formula:C₈H₆Br₄
MDL Number:MFCD03411745
Molecular Weight:421.7500 g/mol
UN:3261
ADR:8 II
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:158.0 to 162.0 deg-C
Specification Test Results
Frequently Bought Together