Crimp caps for crimp neck vials, N 13
Supplier: MACHEREY-NAGEL
Certificates
About this item
The crimp caps with assembled septa are ready-to-use.
- Standard cap with center hole or tamper-evident closure with center tear off cap
- Molded septum with centrical PTFE lamination for very good sealing properties (typically called Pharma-Fix)
Frequently Bought Together