Electronic max/min thermometer
HECH43656010
- Range:−10...+50 °C* / −50...+70 °C**
- Description:Thermometer for indoor/outdoor measurements, cable length approx. 3 m
- Environmentally Preferable:
- Pk:1
Maximum-minimum electronic thermometer used for measuring indoor and outdoor temperature.
- Present temperature/highest and lowest day temperature
- For hanging or standing
- Extra thin cable for easily leading through window seals
- Waterproof sensor for outdoor measurements
* Indoor
** Outdoor
: Incl. 1,5 V AA battery.