Order Entry
Denmark
ContactUsLinkComponent
Rotating mixers, RM 5
Rotating mixers, RM 5
Certificates
undefined
Rotating mixers, RM 5
Specifications
  • Catalog No:
  • HECH40348026
  • 445-2510
  • HECH40349018
  • Model:
  • RM 5
  • RM 5
  • RM 5
  • Shaking motion:
  • Rotating
  • Rotating
  • Rotating
  • Speed:
  • 36 min⁻¹ fixed
  • 36 min⁻¹ fixed
  • 2 - 30 min⁻¹
  • W×D×H:
  • 332×180×95 mm
  • 430×180×95 mm
  • 436×358×105 mm
  • No. of rollers:
  • 5
  • 5
  • 10
  • Weight:
  • 3,2 kg
  • 3,7 kg
  • 3,8 kg
  • Max. load:
  • 2,0 kg
  • 2,0 kg
  • 2,0 kg