Niobium nitride
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:24621-21-4
:
Formula:NNb
MDL Number:MFCD00016239
Density:8.40
Molecular Weight:106.92 g/mol
Storage Temperature:Ambient
EINECS:246-362-5
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together